Anti-HNF1B Rabbit Monoclonal Antibody [clone: EPR6334]

Lieferant: Abcam
AB128912-100
ABCAAB128912-100EA 873 EUR
ABCAAB128912-100
Anti-HNF1B Rabbit Monoclonal Antibody [clone: EPR6334]
Antikörper
Type: Primary
Antigen: HNF1B (HNF1 homeobox B)
Clonality: monoclonal
Clone: EPR6334
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR