Anti-RAB11A Rabbit Monoclonal Antibody [clone: EPR7587]

Lieferant: Abcam
ABCAAB128913-100EA 893 EUR
ABCAAB128913-100
Anti-RAB11A Rabbit Monoclonal Antibody [clone: EPR7587]
Antikörper
Type: Primary
Antigen: RAB11A (RAB11A, member RAS oncogene family)
Clonality: monoclonal
Clone: EPR7587
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR