Anti-SDC1 Rabbit Monoclonal Antibody [clone: EPR6454]
Lieferant: Abcam
ABCAAB128936-100EA 949 EUR
ABCAAB128936-100
Anti-SDC1 Rabbit Monoclonal Antibody [clone: EPR6454]
Antikörper
Type: Primary
Antigen: SDC1 (syndecan 1)
Clonality: monoclonal
Clone: EPR6454
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen