Anti-RGS6 Rabbit Monoclonal Antibody [clone: EPR6342]

Lieferant: Abcam
ABCAAB128943-100EA 777 EUR
ABCAAB128943-100
Anti-RGS6 Rabbit Monoclonal Antibody [clone: EPR6342]
Antikörper
Type: Primary
Antigen: RGS6 (regulator of G-protein signaling 6)
Clonality: monoclonal
Clone: EPR6342
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR