Anti-GTF2F2 Rabbit Monoclonal Antibody [clone: EPR7584]

Lieferant: Abcam
ABCAAB128945-100EA 758 EUR
ABCAAB128945-100
Anti-GTF2F2 Rabbit Monoclonal Antibody [clone: EPR7584]
Antikörper
Type: Primary
Antigen: GTF2F2 (general transcription factor IIF, polypeptide 2, 30kDa)
Clonality: monoclonal
Clone: EPR7584
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR