Anti-EIF3A Rabbit monoclonal antibody unconjugated [clone: EPR5819]

Lieferant: Abcam
AB128996-100
ABCAAB128996-100EA 827 EUR
ABCAAB128996-100
Anti-EIF3A Rabbit monoclonal antibody unconjugated [clone: EPR5819]
Antikörper
Type: Primary
Antigen: EIF3A (eukaryotic translation initiation factor 3, subunit A)
Clonality: monoclonal
Clone: EPR5819
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR