Anti-RAPGEF5 Rabbit Monoclonal Antibody [clone: EPR6882]

Lieferant: Abcam
ABCAAB129008-100EA 787 EUR
ABCAAB129008-100
Anti-RAPGEF5 Rabbit Monoclonal Antibody [clone: EPR6882]
Antikörper
Type: Primary
Antigen: RAPGEF5 (Rap guanine nucleotide exchange factor (GEF) 5)
Clonality: monoclonal
Clone: EPR6882
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR