Anti-PTPN2 Rabbit Monoclonal Antibody [clone: EPR6712]

Lieferant: Abcam
ABCAAB129070-100EA 877 EUR
ABCAAB129070-100
Anti-PTPN2 Rabbit Monoclonal Antibody [clone: EPR6712]
Antikörper
Type: Primary
Antigen: PTPN2 (protein tyrosine phosphatase, non-receptor type 2)
Clonality: monoclonal
Clone: EPR6712
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR