Anti-MFF Rabbit Monoclonal Antibody [clone: EPR7360]

Lieferant: Abcam
ABCAAB129075-100EA 890 EUR
ABCAAB129075-100
Anti-MFF Rabbit Monoclonal Antibody [clone: EPR7360]
Antikörper
Type: Primary
Antigen: MFF (mitochondrial fission factor)
Clonality: monoclonal
Clone: EPR7360
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR