Anti-M Cadherin Rabbit Monoclonal Antibody [clone: EP5756]

Lieferant: Abcam
AB129078-100
ABCAAB129078-100EA 821 EUR
ABCAAB129078-100
Anti-M Cadherin Rabbit Monoclonal Antibody [clone: EP5756]
Antikörper
Type: Primary
Antigen: M Cadherin
Clonality: monoclonal
Clone: EP5756
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR