Anti-GNB2L1 Rabbit monoclonal antibody unconjugated [clone: EPR7388]

Lieferant: Abcam
ABCAAB129084-100EA 831 EUR
ABCAAB129084-100
Anti-GNB2L1 Rabbit monoclonal antibody unconjugated [clone: EPR7388]
Antikörper
Type: Primary
Antigen: GNB2L1 (guanine nucleotide binding protein (G protein), beta polypeptide 2-like 1)
Clonality: monoclonal
Clone: EPR7388
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR