Anti-ERCC1 Rabbit Monoclonal Antibody [clone: EPR7062]
Lieferant: Abcam
ABCAAB129093-100EA 855 EUR
ABCAAB129093-100
Anti-ERCC1 Rabbit Monoclonal Antibody [clone: EPR7062]
Antikörper
Type: Primary
Antigen: ERCC1 (excision repair cross-complementation group 1)
Clonality: monoclonal
Clone: EPR7062
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen