Anti-CS Rabbit monoclonal antibody unconjugated [clone: EPR8067]
Lieferant: Abcam
ABCAAB129095-100EA 908 EUR
ABCAAB129095-100
Anti-CS Rabbit monoclonal antibody unconjugated [clone: EPR8067]
Antikörper
Type: Primary
Antigen: CS (citrate synthase)
Clonality: monoclonal
Clone: EPR8067
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen