Anti-SP3 Rabbit Monoclonal Antibody [clone: EPR6806]

Lieferant: Abcam
ABCAAB129099-100EA 791 EUR
ABCAAB129099-100
Anti-SP3 Rabbit Monoclonal Antibody [clone: EPR6806]
Antikörper
Type: Primary
Antigen: SP3 (Sp3 transcription factor)
Clonality: monoclonal
Clone: EPR6806
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR