Anti-CDC123 Rabbit Monoclonal Antibody [clone: EP5861]
Lieferant: Abcam
ABCAAB129101-100EA 761 EUR
ABCAAB129101-100
Anti-CDC123 Rabbit Monoclonal Antibody [clone: EP5861]
Antikörper
Type: Primary
Antigen: CDC123 (cell division cycle 123)
Clonality: monoclonal
Clone: EP5861
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen