Anti-EDNRB Rabbit monoclonal antibody unconjugated [clone: EPR7013]

Lieferant: Abcam
AB129102-100
ABCAAB129102-100EA 862 EUR
ABCAAB129102-100
Anti-EDNRB Rabbit monoclonal antibody unconjugated [clone: EPR7013]
Antikörper
Type: Primary
Antigen: EDNRB (endothelin receptor type B)
Clonality: monoclonal
Clone: EPR7013
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR