Anti-SRSF1 Rabbit Monoclonal Antibody [clone: EPR8239]
Lieferant: Abcam
ABCAAB129108-100EA 897 EUR
ABCAAB129108-100
Anti-SRSF1 Rabbit Monoclonal Antibody [clone: EPR8239]
Antikörper
Type: Primary
Antigen: SRSF1 (serine/arginine-rich splicing factor 1)
Clonality: monoclonal
Clone: EPR8239
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen