Anti-MFN1 Rabbit Monoclonal Antibody [clone: EPR7960]

Lieferant: Abcam
AB129154-100
ABCAAB129154-100EA 890 EUR
ABCAAB129154-100
Anti-MFN1 Rabbit Monoclonal Antibody [clone: EPR7960]
Antikörper
Type: Primary
Antigen: MFN1 (mitofusin 1)
Clonality: monoclonal
Clone: EPR7960
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR