Anti-Osteogenic Growth Peptide Rabbit Monoclonal Antibody [clone: EPR5897]
Lieferant: Abcam
ABCAAB129156-100EA 855 EUR
ABCAAB129156-100
Anti-Osteogenic Growth Peptide Rabbit Monoclonal Antibody [clone: EPR5897]
Antikörper
Type: Primary
Antigen: Osteogenic Growth Peptide
Clonality: monoclonal
Clone: EPR5897
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen