Anti-NUDT5 Rabbit Monoclonal Antibody [clone: EPR7735]

Lieferant: Abcam
ABCAAB129163-100EA 781 EUR
ABCAAB129163-100
Anti-NUDT5 Rabbit Monoclonal Antibody [clone: EPR7735]
Antikörper
Type: Primary
Antigen: NUDT5 (nudix (nucleoside diphosphate linked moiety X)-type motif 5)
Clonality: monoclonal
Clone: EPR7735
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR