Anti-MED4 Rabbit Monoclonal Antibody [clone: EPR7816]

Lieferant: Abcam
AB129170-100
ABCAAB129170-100EA 817 EUR
ABCAAB129170-100
Anti-MED4 Rabbit Monoclonal Antibody [clone: EPR7816]
Antikörper
Type: Primary
Antigen: MED4 (mediator complex subunit 4)
Clonality: monoclonal
Clone: EPR7816
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR