Anti-THEMIS Rabbit Monoclonal Antibody [clone: EPR7354]

Lieferant: Abcam
ABCAAB129174-100EA 758 EUR
ABCAAB129174-100
Anti-THEMIS Rabbit Monoclonal Antibody [clone: EPR7354]
Antikörper
Type: Primary
Antigen: THEMIS (thymocyte selection associated)
Clonality: monoclonal
Clone: EPR7354
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR