Anti-BANF1 Rabbit monoclonal antibody unconjugated [clone: EPR7668]

Lieferant: Abcam
ABCAAB129184-100EA 778 EUR
ABCAAB129184-100
Anti-BANF1 Rabbit monoclonal antibody unconjugated [clone: EPR7668]
Antikörper
Type: Primary
Antigen: BANF1 (barrier to autointegration factor 1)
Clonality: monoclonal
Clone: EPR7668
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR