Anti-BAD Rabbit monoclonal antibody unconjugated [clone: EPR1891]

Lieferant: Abcam
AB129192-100
ABCAAB129192-100EA 827 EUR
ABCAAB129192-100
Anti-BAD Rabbit monoclonal antibody unconjugated [clone: EPR1891]
Antikörper
Type: Primary
Antigen: BAD (BCL2-associated agonist of cell death)
Clonality: monoclonal
Clone: EPR1891
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR