Anti-NCK1 Mouse Monoclonal Antibody [clone: 5B7]

Lieferant: Abcam
AB129367-100
ABCAAB129367-100EA 906 EUR
ABCAAB129367-100
Anti-NCK1 Mouse Monoclonal Antibody [clone: 5B7]
Antikörper
Type: Primary
Antigen: NCK1 (NCK adaptor protein 1)
Clonality: monoclonal
Clone: 5B7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR