Anti-GHRL Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB129383-100EA 840 EUR
ABCAAB129383-100
Anti-GHRL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GHRL (ghrelin/obestatin prepropeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR