Anti-CCM2 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB129386-100EA 797 EUR
ABCAAB129386-100
Anti-CCM2 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CCM2 (cerebral cavernous malformation 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR