Anti-MAP2K1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB129431-100EA 852 EUR
ABCAAB129431-100
Anti-MAP2K1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR