Anti-NDUFA4 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB129752-100EA 840 EUR
ABCAAB129752-100
Anti-NDUFA4 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NDUFA4 (NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4, 9kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR