Anti-RPL3L Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB129753-100EA 797 EUR
ABCAAB129753-100
Anti-RPL3L Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RPL3L (ribosomal protein L3-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR