Anti-RAC1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB129758-50EA 840 EUR
ABCAAB129758-50
Anti-RAC1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAC1 (ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR