Anti-TAS2R39 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB130074-100
ABCAAB130074-100EA 882 EUR
ABCAAB130074-100
Anti-TAS2R39 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TAS2R39 (taste receptor, type 2, member 39)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR