Anti-FAT4 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB130076-100EA 840 EUR
ABCAAB130076-100
Anti-FAT4 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: FAT4 (FAT atypical cadherin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR