Anti-IL11 Mouse Monoclonal Antibody [clone: 6D9A1]

Lieferant: Abcam
AB130083-100
ABCAAB130083-100EA 882 EUR
ABCAAB130083-100
Anti-IL11 Mouse Monoclonal Antibody [clone: 6D9A1]
Antikörper
Type: Primary
Antigen: IL11 (interleukin 11)
Clonality: monoclonal
Clone: 6D9A1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR