Anti-DCHS1 Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB130092-100EA 835 EUR
ABCAAB130092-100
Anti-DCHS1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: DCHS1 (dachsous cadherin-related 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen