Anti-BMPR2 Mouse Monoclonal Antibody [clone: 1F12]

Lieferant: Abcam
ABCAAB130206-100EA 869 EUR
ABCAAB130206-100
Anti-BMPR2 Mouse Monoclonal Antibody [clone: 1F12]
Antikörper
Type: Primary
Antigen: BMPR2 (bone morphogenetic protein receptor, type II (serine/threonine kinase))
Clonality: monoclonal
Clone: 1F12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR