Anti-CD79B Mouse Monoclonal Antibody [clone: CB3-1]

Lieferant: Abcam
ABCAAB130422-100EA 930 EUR
ABCAAB130422-100
Anti-CD79B Mouse Monoclonal Antibody [clone: CB3-1]
Antikörper
Type: Primary
Antigen: CD79B (CD79b molecule, immunoglobulin-associated beta)
Clonality: monoclonal
Clone: CB3-1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR