Anti-IgM Goat Polyclonal Antibody (APC (Allophycocyanin))

Lieferant: Abcam
AB130780-125
ABCAAB130780-125EA 413 EUR
ABCAAB130780-125
Anti-IgM Goat Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Type: Secondary
Antigen: IgM (Mouse) (mouse IgM)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR