Anti-IgG3 Goat Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)

Lieferant: Abcam
ABCAAB130794-125EA 393 EUR
ABCAAB130794-125
Anti-IgG3 Goat Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)
Antikörper
Type: Secondary
Antigen: IgG3 (Mouse) (mouse IgG3)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR