Anti-IgG Goat Polyclonal Antibody (Spectral Red®)

Lieferant: Abcam
ABCAAB130802-125EA 393 EUR
ABCAAB130802-125
Anti-IgG Goat Polyclonal Antibody (Spectral Red®)
Antikörper
Type: Secondary
Antigen: IgG (Rat) (rat IgG)
Clonality: polyclonal
Clone:
Conjugation: Spectral Red®
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR