Anti-FCGR2B Rabbit monoclonal antibody unconjugated [clone: EP6243]

Lieferant: Abcam
ABCAAB131051-100EA 856 EUR
ABCAAB131051-100
Anti-FCGR2B Rabbit monoclonal antibody unconjugated [clone: EP6243]
Antikörper
Type: Primary
Antigen: FCGR2B (Fc fragment of IgG, low affinity IIb, receptor (CD32))
Clonality: monoclonal
Clone: EP6243
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR