Anti-NOX1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB131088-100EA 840 EUR
ABCAAB131088-100
Anti-NOX1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NOX1 (NADPH oxidase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR