Anti-GSK3A Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB131112-100EA 859 EUR
ABCAAB131112-100
Anti-GSK3A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GSK3A (glycogen synthase kinase 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR