Anti-TOP1 Rabbit Monoclonal Antibody [clone: EPR5376]

Lieferant: Abcam
ABCAAB131166-100EA 880 EUR
ABCAAB131166-100
Anti-TOP1 Rabbit Monoclonal Antibody [clone: EPR5376]
Antikörper
Type: Primary
Antigen: TOP1 (topoisomerase (DNA) I)
Clonality: monoclonal
Clone: EPR5376
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR