Anti-ATP7B Rabbit monoclonal antibody unconjugated [clone: EPR6793]

Lieferant: Abcam
ABCAAB131208-100EA 847 EUR
ABCAAB131208-100
Anti-ATP7B Rabbit monoclonal antibody unconjugated [clone: EPR6793]
Antikörper
Type: Primary
Antigen: ATP7B (ATPase, Cu++ transporting, beta polypeptide)
Clonality: monoclonal
Clone: EPR6793
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR