Anti-GBP1 Rabbit monoclonal antibody unconjugated [clone: EPR8285]

Lieferant: Abcam
AB131255-100
ABCAAB131255-100EA 898 EUR
ABCAAB131255-100
Anti-GBP1 Rabbit monoclonal antibody unconjugated [clone: EPR8285]
Antikörper
Type: Primary
Antigen: GBP1 (guanylate binding protein 1, interferon-inducible)
Clonality: monoclonal
Clone: EPR8285
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR