Anti-Neutrophil Elastase Rabbit Monoclonal Antibody [clone: EPR7479]

Lieferant: Abcam
AB131260-100
ABCAAB131260-100EA 991 EUR
ABCAAB131260-100
Anti-Neutrophil Elastase Rabbit Monoclonal Antibody [clone: EPR7479]
Antikörper
Type: Primary
Antigen: Neutrophil Elastase
Clonality: monoclonal
Clone: EPR7479
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR