Anti-CD2 Rabbit Monoclonal Antibody [clone: EPR6451]

Lieferant: Abcam
ABCAAB131276-100EA 854 EUR
ABCAAB131276-100
Anti-CD2 Rabbit Monoclonal Antibody [clone: EPR6451]
Antikörper
Type: Primary
Antigen: CD2 (CD2 molecule)
Clonality: monoclonal
Clone: EPR6451
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR