Anti-USP39 Rabbit Monoclonal Antibody [clone: EPR8684]

Lieferant: Abcam
AB131332-100
ABCAAB131332-100EA 819 EUR
ABCAAB131332-100
Anti-USP39 Rabbit Monoclonal Antibody [clone: EPR8684]
Antikörper
Type: Primary
Antigen: USP39 (ubiquitin specific peptidase 39)
Clonality: monoclonal
Clone: EPR8684
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR