Anti-CAMK2A Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB131468-100EA 840 EUR
ABCAAB131468-100
Anti-CAMK2A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CAMK2A (calcium/calmodulin-dependent protein kinase II alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR